ENA321763287 Enamine

2-(1-chloroethyl)-5-ethyl-1,3,4-oxadiazole

  • MDL number MFCD09041356
  • Cas Number : 926214-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74404
Availability details for
Quantity
  • Availability
  • Add to Cart