ENA321763295 Enamine

pentane-1-sulfonamide

  • MDL number MFCD09800672
  • Cas Number : 52960-14-2
  • Molecular Weight : 151.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74910
Availability details for
Quantity
  • Availability
  • Add to Cart