ENA321763300 Enamine

N1-phenylpropane-1,2-diamine

  • MDL number MFCD12796450
  • Cas Number : 6499-72-5
  • Molecular Weight : 150.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74916
Availability details for
Quantity
  • Availability
  • Add to Cart