ENA321763301 Enamine

{3-[(oxan-4-yloxy)methyl]phenyl}methanamine

  • MDL number MFCD14614909
  • Cas Number : 1251264-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74919
Availability details for
Quantity
  • Availability
  • Add to Cart