ENA321763306 Enamine

2-amino-5-bromo-N-methylpyridine-3-sulfonamide

  • MDL number MFCD16688567
  • Cas Number : 1086063-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74410

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74410
Availability details for
Quantity
  • Availability
  • Add to Cart