ENA321763309 Enamine

2-{[(2,4-dimethoxyphenyl)methyl]amino}ethan-1-ol

  • MDL number MFCD07407284
  • Cas Number : 40171-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74931

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-74931
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart