ENA321763310 Enamine

3-methyl-3,4-dihydro-2H-1-benzothiopyran-4-one

  • MDL number MFCD12132764
  • Cas Number : 771-17-5
  • Molecular Weight : 178.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74932
Availability details for
Quantity
  • Availability
  • Add to Cart