ENA321763313 Enamine

N-[(4-fluorophenyl)methyl]-2-hydroxyacetamide

  • MDL number MFCD12138145
  • Cas Number : 1153759-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74412
Availability details for
Quantity
  • Availability
  • Add to Cart