ENA321763317 Enamine

3-(2,6-dichlorophenyl)propanenitrile

  • MDL number MFCD09744610
  • Cas Number : 61698-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40...42
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74944

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74944
Availability details for
Quantity
  • Availability
  • Add to Cart