ENA321763318 Enamine

4-[(5-chloropyridin-2-yl)oxy]benzoic acid

  • MDL number MFCD13480743
  • Cas Number : 289044-48-0
  • Molecular Weight : 249.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74945
Availability details for
Quantity
  • Availability
  • Add to Cart