ENA321763320 Enamine

(1S)-1-[3-(trifluoromethyl)phenyl]ethan-1-ol

  • MDL number MFCD03093004
  • Cas Number : 96789-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74313

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74313
Availability details for
Quantity
  • Availability
  • Add to Cart