ENA321763323 Enamine

{1-[(4,5-dimethyl-1,3-oxazol-2-yl)methyl]piperidin-4-yl}methanamine

  • MDL number MFCD16169516
  • Cas Number : 1249949-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74952

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74952
Availability details for
Quantity
  • Availability
  • Add to Cart