ENA321763324 Enamine

5-(3-bromophenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD01590338
  • Cas Number : 108656-65-1
  • Molecular Weight : 256.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74416
Availability details for
Quantity
  • Availability
  • Add to Cart