ENA321763329 Enamine

2-chloro-6-(dimethylamino)pyridine-4-carboxylic acid

  • MDL number MFCD11214889
  • Cas Number : 56836-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74977

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74977
Availability details for
Quantity
  • Availability
  • Add to Cart