ENA321763330 Enamine

1-(1,4-dimethylpiperazin-2-yl)methanamine

  • MDL number MFCD13151976
  • Cas Number : 131922-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74417
Availability details for
Quantity
  • Availability
  • Add to Cart