ENA321763331 Enamine

3,5-dimethyl-2-phenylmorpholine

  • MDL number MFCD11653056
  • Cas Number : 1218345-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74418
Availability details for
Quantity
  • Availability
  • Add to Cart