ENA321763332 Enamine

3-[(pyrrolidin-1-yl)carbonyl]piperidine hydrochloride

  • MDL number MFCD11520873
  • Cas Number : 937724-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187...189
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74980

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74980
Availability details for
Quantity
  • Availability
  • Add to Cart