ENA321763334 Enamine

1-(2,5-dimethylthiophen-3-yl)-2-methylpropan-1-amine

  • MDL number MFCD10693724
  • Cas Number : 1152536-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74982
Availability details for
Quantity
  • Availability
  • Add to Cart