ENA321763342 Enamine

2-[3-(trifluoromethyl)phenyl]-1H-imidazole

  • MDL number MFCD08668838
  • Cas Number : 27423-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74995
Availability details for
Quantity
  • Availability
  • Add to Cart