ENA321763343 Enamine

4-(2H-1,3-benzodioxol-5-yl)-3-(furan-3-yl)-1,2-oxazol-5-amine

  • MDL number MFCD12541301
  • Cas Number : 1096939-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74996

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74996
Availability details for
Quantity
  • Availability
  • Add to Cart