ENA321763345 Enamine

1-(2-aminophenyl)pyrrolidine-2,5-dione

  • MDL number MFCD00022591
  • Cas Number : 1012-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231...233
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74999

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-74999
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart