ENA321763347 Enamine

2-methyl-1-(3-methyl-1,2,4-oxadiazol-5-yl)propan-1-amine

  • MDL number MFCD11122292
  • Cas Number : 1041603-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75001
Availability details for
Quantity
  • Availability
  • Add to Cart