ENA321763351 Enamine

7-chloro-8-fluoro-4-hydroxyquinoline-3-carboxylic acid

  • MDL number MFCD09861319
  • Cas Number : 1019015-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75005

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75005
Availability details for
Quantity
  • Availability
  • Add to Cart