ENA321763354 Enamine

5-methyl-2-(2-oxopyrrolidin-1-yl)benzoic acid

  • MDL number MFCD11127759
  • Cas Number : 1019394-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75008
Availability details for
Quantity
  • Availability
  • Add to Cart