ENA321763357 Enamine

3-methyl-3-(thiophen-2-yl)butanoic acid

  • MDL number MFCD09837229
  • Cas Number : 937640-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74316

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74316
Availability details for
Quantity
  • Availability
  • Add to Cart