ENA321763360 Enamine

2-[(2-amino-4-methylphenyl)sulfanyl]-N-(2-methylphenyl)acetamide

  • MDL number MFCD12575311
  • Cas Number : 1095624-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74436
Availability details for
Quantity
  • Availability
  • Add to Cart