ENA321763362 Enamine

4-chloro-2-[(oxolan-3-ylmethyl)amino]benzonitrile

  • MDL number MFCD12118994
  • Cas Number : 1153004-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74437

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74437
Availability details for
Quantity
  • Availability
  • Add to Cart