ENA321763364 Enamine

2-amino-2,4-dimethylpentanamide

  • MDL number MFCD12151846
  • Cas Number : 113509-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75027
Availability details for
Quantity
  • Availability
  • Add to Cart