ENA321763370 Enamine

3-(4-nitrophenyl)adamantan-1-ol

  • Cas Number : 42009-76-7
  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74440
Availability details for
Quantity
  • Availability
  • Add to Cart