ENA321763372 Enamine

1-amino-2,3-dimethylbutan-2-ol

  • MDL number MFCD12169421
  • Cas Number : 1154150-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75043
Availability details for
Quantity
  • Availability
  • Add to Cart