ENA321763373 Enamine

8-ethyl-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD09732239
  • Cas Number : 103858-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75044
Availability details for
Quantity
  • Availability
  • Add to Cart