ENA321763375 Enamine

3-methylbutane-1-sulfonamide

  • MDL number MFCD12547174
  • Cas Number : 98138-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74442
Availability details for
Quantity
  • Availability
  • Add to Cart