ENA321763384 Enamine

5-(ethanesulfonyl)-2-fluorobenzoic acid

  • MDL number MFCD11179894
  • Cas Number : 898258-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75075
Availability details for
Quantity
  • Availability
  • Add to Cart