ENA321763387 Enamine

3-[(2,5-difluorophenyl)sulfanyl]propanoic acid

  • MDL number MFCD09050963
  • Cas Number : 926232-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74295
Availability details for
Quantity
  • Availability
  • Add to Cart