ENA321763390 Enamine

1-methyl-1H,4H,5H,6H,7H-imidazo[4,5-c]pyridine dihydrochloride

  • MDL number MFCD18785463
  • Cas Number : 485402-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239...241
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74447

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74447
Availability details for
Quantity
  • Availability
  • Add to Cart