ENA321763394 Enamine

[1-(2,2,2-trifluoroethyl)piperidin-4-yl]methanol

  • MDL number MFCD16153513
  • Cas Number : 831169-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75088
Availability details for
Quantity
  • Availability
  • Add to Cart