ENA321763395 Enamine

1-(2-chlorophenyl)-2-(4-fluorophenyl)ethan-1-one

  • MDL number MFCD12863030
  • Cas Number : 137101-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75089
Availability details for
Quantity
  • Availability
  • Add to Cart