ENA321763397 Enamine

2-amino-N-phenylpropanamide hydrochloride

  • MDL number MFCD13562609
  • Cas Number : 153973-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 255...257
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75095

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75095
Availability details for
Quantity
  • Availability
  • Add to Cart