ENA321763399 Enamine

4-ethoxy-3-methylbenzoic acid

  • MDL number MFCD13920122
  • Cas Number : 92315-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198...200
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75097

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75097
Availability details for
Quantity
  • Availability
  • Add to Cart