ENA321763403 Enamine

2-[methyl(propan-2-yl)amino]benzoic acid

  • MDL number MFCD11156542
  • Cas Number : 1021244-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75102
Availability details for
Quantity
  • Availability
  • Add to Cart