ENA321763405 Enamine

4-(thiophen-2-yl)butanehydrazide

  • MDL number MFCD09808386
  • Cas Number : 1016494-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75104

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75104
Availability details for
Quantity
  • Availability
  • Add to Cart