ENA321763410 Enamine

tert-butyl N-(1-cyanopropan-2-yl)-N-methylcarbamate

  • MDL number MFCD18297433
  • Cas Number : 1311315-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75111

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75111
Availability details for
Quantity
  • Availability
  • Add to Cart