ENA321763416 Enamine

2-(4-aminopiperidin-1-yl)benzamide

  • MDL number MFCD12137450
  • Cas Number : 1039022-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75123

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-75123
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart