ENA321763425 Enamine

4-chloro-3-[(2-methyl-1H-1,3-benzodiazol-1-yl)methyl]aniline

  • MDL number MFCD12655680
  • Cas Number : 1097805-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74456

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74456
Availability details for
Quantity
  • Availability
  • Add to Cart