ENA321763426 Enamine

3-(oxan-4-yl)prop-2-enoic acid

  • MDL number MFCD16745937
  • Cas Number : 1278963-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75140

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75140
Availability details for
Quantity
  • Availability
  • Add to Cart