ENA321763428 Enamine

5-ethyl-5-(propan-2-yl)imidazolidine-2,4-dione

  • MDL number MFCD12807236
  • Cas Number : 98492-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75142

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75142
Availability details for
Quantity
  • Availability
  • Add to Cart