ENA321763432 Enamine

[1-benzothiophen-3-yl(pyridin-3-yl)methyl](ethyl)amine

  • MDL number MFCD12474159
  • Cas Number : 1154227-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75146

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75146
Availability details for
Quantity
  • Availability
  • Add to Cart