ENA321763433 Enamine

1-(2-chlorobenzoyl)piperidin-3-ol

  • MDL number MFCD11738633
  • Cas Number : 1090598-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75147
Availability details for
Quantity
  • Availability
  • Add to Cart