ENA321763434 Enamine

6,7-difluoro-3,4-dihydroquinazolin-4-one

  • MDL number MFCD18838685
  • Cas Number : 205259-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75150
Availability details for
Quantity
  • Availability
  • Add to Cart