ENA321763438 Enamine

[(4-bromo-2-fluorophenyl)methyl][2-(4-methylpiperazin-1-yl)ethyl]amine

  • MDL number MFCD13235091
  • Cas Number : 1183009-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75155

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75155
Availability details for
Quantity
  • Availability
  • Add to Cart